Learn More
1,4-Dichloro-2-butyne, TRC
Product Code. 30386621
Change view
Click to view available options
Quantity:
50 g
Unit Size:
50g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30386621 50 g 50g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30386621 Supplier Toronto Research Chemicals Supplier No. TRCD43237050G
 Restricted Item
Estimated Shipment: 23-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,4-Dichloro-2-butyne
CAS 821-10-3
Molecular Formula C4H4Cl2
Quantity 50 g
Synonym 1,4-Dichloro-2-butyne, 1,4-Dichlorobutyne, Bis(chloromethyl)acetylene, NSC 30603
InChI Formula InChI=1S/C4H4Cl2/c5-3-1-2-4-6/h3-4H2
SMILES ClCC#CCCl
Molecular Weight (g/mol) 122.98