missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,4-Dibromobutane-1,1,4,4-d4, CDN
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.5g
1g
Specifications
Specifications
| Chemical Name or Material | 1,4-Dibromobutane-1,1,4,4-D4 |
| CAS | 36684-45-4 |
| Molecular Formula | C4 D4 H4 Br2 |
| Quantity | 1 g |
| Synonym | Butane-1,1,4,4-d4, 1,4-dibromo- (9 CI, ACI), 1,4-Dibromobutane-1,1,4,4-d4 (ACI), 1,1,4,4-Tetradeutero-1,4-dibromobutane, 1,4-Dibromobutane-d4, 1,4-Dibromobutane-1,1,4,4-D4 |
| InChI Formula | InChI=1 S/C4H8Br2/c5-3-1-2-4-6/h1-4H2/i3D2,4D2 |
| SMILES | [2 H]C([2 H])(Br)CCC([2 H])([2 H])Br |
| IUPAC Name | 1,4-dibromo-1,1,4,4-tetradeuteriobutane |
| Molecular Weight (g/mol) | 219.939 |
| Formula Weight | 217.9244 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction