Learn More
1,4-Cyclohexanediamine, TRC
Specifications
Specifications
| CAS | 3114-70-3 |
| Molecular Formula | C6H14N2 |
| Quantity | 100 mg |
| Synonym | 1,4-Cyclohexanediamine (6CI, 7CI, 8CI, 9CI, ACI), 1,4-Cyclohexylenediamine, 1,4-Diaminocyclohexane, 4-Aminocyclohexylamine, Hexahydro-1,4-phenylenediamine, Cyclohexane-1,4-diamine |
| InChI Formula | InChI=1S/C6H14N2/c7-5-1-2-6(8)4-3-5/h5-6H,1-4,7-8H2 |
| SMILES | NC1CCC(N)CC1 |
| IUPAC Name | cyclohexane-1,4-diamine |
| Molecular Weight (g/mol) | 114.19 |
| Formula Weight | 114.1157 |
| Chemical Name or Material | 1,4-Cyclohexanediamine |