Learn More
1,4-Butanediol Terephthalate, TRC
Specifications
Specifications
| Chemical Name or Material | 1,4-Butanediol Terephthalate |
| CAS | 29278-69-1 |
| Molecular Formula | C20H18O8 |
| Quantity | 250 mg |
| Synonym | 4,4'-((Butane-1,4-diylbis(oxy))bis(carbonyl))dibenzoic Acid, 1,4-Benzenedicarboxylic Acid 1,4-Butanediyl Ester, 1,4-Butanediol bis(Hydrogen Terephthalate), Terephthalic Acid Tetramethylene Ester |
| InChI Formula | InChI=1S/C20H18O8/c21-17(22)13-3-7-15(8-4-13)19(25)27-11-1-2-12-28-20(26)16-9-5-14(6-10-16)18(23)24/h3-10H,1-2,11-12H2,(H,21,22)(H,23,24) |
| SMILES | OC(=O)c1ccc(cc1)C(=O)OCCCCOC(=O)c2ccc(cc2)C(=O)O |
| IUPAC Name | 4-[4-(4-carboxybenzoyl)oxybutoxycarbonyl]benzoic acid |
| Molecular Weight (g/mol) | 386.35 |
| Formula Weight | 386.1002 |