Learn More
1,4-Butane-d8-diamine Dihydrochloride, TRC
Product Code. 30359810
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30359810 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30359810 Supplier Toronto Research Chemicals Supplier No. TRCD416021100MG
 Restricted Item
Estimated Shipment: 11-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,4-Diaminobutane-D8 Dihydrochloride
CAS 284665-22-1
Molecular Formula C4 D8 H4 N2 . 2 Cl H
Quantity 100 mg
Synonym 1,4-Butane-1,1,2,2,3,3,4,4-d8-diamine, dihydrochloride (9CI), 1,4-Butane-d8-diamine dihydrochloride, 1,4-Diaminobutane D8 dihydrochloride, 1,4-Diaminobutane-D8 dihydrochloride, 1,4-Butane-1,1,2,2,3,3,4,4-d8-diamine dihydrochloride, 1,4-Butane-d8-diamine d
InChI Formula InChI=1S/C4H12N2.2ClH/c5-3-1-2-4-6;;/h1-6H2;2*1H/i1D2,2D2,3D2,4D2;;
SMILES Cl.Cl.[2H]C([2H])(N)C([2H])([2H])C([2H])([2H])C([2H])([2H])N
IUPAC Name 1,1,2,2,3,3,4,4-octadeuteriobutane-1,4-diamine;dihydrochloride
Molecular Weight (g/mol) 169.123
Formula Weight 168.104