missing translation for 'onlineSavingsMsg'
Learn More

1,4-Butane-2,2,3,3-d4-diamine 2HCl, TRC

Product Code. 30365908
Change view
Click to view available options
Quantity:
1 mg
2 mg
10 mg
Unit Size:
10mg
1mg
2mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30365908 10 mg 10mg
30367832 1 mg 1mg
30350394 2 mg 2mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30365908 Supplier Toronto Research Chemicals Supplier No. TRCD41602210MG
 Restricted Item
Estimated Shipment: 19-08-2026
Add to basket
Add to basket
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 88972-24-1
Molecular Formula C4H10D4Cl2N2
Molecular Weight (g/mol) 165.09
Synonym 1,4-Butane-2,2,3,3-d4-diamine, dihydrochloride (9CI), 1,4-Butane-2,2,3,3-d4-diamine 2HCl, 1,4-Butane-2,2,3,3-d4-diamine dihydrochloride, 1,4-Diaminobutane-2,2,3,3-D4 dihydrochloride, 1,4-Diaminobutane-d4 dihydrochloride, Putrescine-d4 dihydrochloride
IUPAC Name butane-2,2,3,3-d4-1,4-diamine hydrochloride
SMILES C(C(CN)([2H])[2H])(CN)([2H])[2H].Cl

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C4H12N2.ClH/c5-3-1-2-4-6;/h1-6H2;1H/i1D2,2D2;
Formula Weight 164.08
Chemical Name or Material 1,4-Diaminobutane-2,2,3,3-D4 Dihydrochloride
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.