Learn More
1-(4-Bromo-3-methylphenyl)ethanone, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(4-Bromo-3-methylphenyl)ethanone |
| CAS | 37074-40-1 |
| Molecular Formula | C9H9BrO |
| Quantity | 1 g |
| InChI Formula | InChI=1S/C9H9BrO/c1-6-5-8(7(2)11)3-4-9(6)10/h3-5H,1-2H3 |
| SMILES | CC1=C(C=CC(=C1)C(=O)C)Br |
| IUPAC Name | 1-(4-bromo-3-methylphenyl)ethanone |
| Molecular Weight (g/mol) | 213.07 |
| Formula Weight | 211.9837 |