missing translation for 'onlineSavingsMsg'
Learn More

1-(4-Biphenylyl)ethylamine, 97%, Thermo Scientific™

Product Code. 15474558
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15474558 5 g 5g
15464558 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15474558 Supplier Thermo Scientific Alfa Aesar Supplier No. H58237.06
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.

Applications
It is used as pharmaceutical intermediate.

Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 86217-82-5
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.281
MDL Number MFCD02667819
InChI Key QVLZRPPCCDKMPO-UHFFFAOYSA-N
Synonym 1-4-phenylphenyl ethanamine, 1-1,1'-biphenyl-4-yl ethanamine, chembl19081, 1,1'-biphenyl-4-methanamine,a-methyl, 1-4-phenylphenyl ethylamine, 4-1-aminoethyl biphenyl, 1-4-biphenylyl ethylamine, 1-biphenyl-4-yl-ethylamine
PubChem CID 2771754
IUPAC Name 1-(4-phenylphenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)C2=CC=CC=C2)N

Specifications

Quantity 5 g
Formula Weight 197.28
Percent Purity 97%
Chemical Name or Material 1-(4-Biphenylyl)ethylamine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.