Learn More
1-(4-Biphenylyl)ethylamine, 97%, Thermo Scientific™
Product Code. 15464558
Change view
Click to view available options
Quantity:
1 g
5 g
Unit Size:
1g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15464558 1 g 1g
15474558 5 g 5g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15464558 Supplier Thermo Scientific Alfa Aesar Supplier No. H58237.03
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
Applications
It is used as pharmaceutical intermediate.
Notes
Store at room temperature. Incompatible with strong oxidizing agents and acids.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 86217-82-5
Molecular Formula C14H15N
Molecular Weight (g/mol) 197.281
MDL Number MFCD02667819
InChI Key QVLZRPPCCDKMPO-UHFFFAOYSA-N
Synonym 1-4-phenylphenyl ethanamine, 1-1,1'-biphenyl-4-yl ethanamine, chembl19081, 1,1'-biphenyl-4-methanamine,a-methyl, 1-4-phenylphenyl ethylamine, 4-1-aminoethyl biphenyl, 1-4-biphenylyl ethylamine, 1-biphenyl-4-yl-ethylamine
PubChem CID 2771754
IUPAC Name 1-(4-phenylphenyl)ethanamine
SMILES CC(C1=CC=C(C=C1)C2=CC=CC=C2)N
Specifications
Quantity 1 g
Formula Weight 197.28
Percent Purity 97%
Chemical Name or Material 1-(4-Biphenylyl)ethylamine