Learn More
1-(4-Aza-8-hydroxy-6-oxo)oct-2-en-1-oylimidazole(mixture E/Z), TRC
Specifications
Specifications
| Chemical Name or Material | 1-(4-Aza-8-hydroxy-6-oxo)oct-2-en-1-oylimidazole(mixture E/Z) |
| CAS | 89186-34-5 |
| Molecular Formula | C10H13N3O3 |
| Quantity | 5 mg |
| Synonym | 4-Hydroxy-1-[[3-(1H-imidazol-1-yl)-3-oxo-1-propen-1-yl]amino]-2-butanone, 1-[3-[(4-Hydroxy-2-oxobutyl)amino]-1-oxo-2-propenyl]-1H-imidazole |
| InChI Formula | InChI=1S/C10H13N3O3/c14-6-2-9(15)7-11-3-1-10(16)13-5-4-12-8-13/h1,3-5,8,11,14H,2,6-7H2 |
| SMILES | O=C(N1C=NC=C1)C=CNCC(CCO)=O |
| IUPAC Name | 1-((3-(1H-imidazol-1-yl)-3-oxoprop-1-en-1-yl)amino)-4-hydroxybutan-2-one |
| Molecular Weight (g/mol) | 223.23 |
| Formula Weight | 223.0957 |