missing translation for 'onlineSavingsMsg'
Learn More

1-[4-(1H-Indol-3-yl)piperidino]ethan-1-one, 97%, Thermo Scientific™

Product Code. 10456482
Click to view available options
Quantity:
250 mg
1 g
5 g
Unit Size:
1g
250mg
5g
This item is not returnable. View return policy

Product Code. 10456482

Brand: Thermo Scientific Maybridge RH01995DA

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Chemical Identifiers

CAS 30030-83-2
Molecular Formula C15H18N2O
Molecular Weight (g/mol) 242.322
MDL Number MFCD01764639
InChI Key PRFIVCWVSORYRV-UHFFFAOYSA-N
Synonym 1-4-1h-indol-3-yl piperidino ethan-1-one, 1-4-1h-indol-3-yl piperidin-1-yl ethanone, maybridge3_005459, 3-n-acetyl-4-piperidyl indole, 1-acetyl-4-indol-3-ylpiperidine, 3-1-acetyl-4-piperidyl indole, 1-acetyl-4-1h-indol-3-yl piperidine, 3-1-acetylpiperidin-4-yl-1h-indole, 1-4-1h-indol-3-yl-piperidin-1-yl-ethanone
PubChem CID 2728359
IUPAC Name 1-[4-(1H-indol-3-yl)piperidin-1-yl]ethanone
SMILES CC(=O)N1CCC(CC1)C2=CNC3=CC=CC=C32

Specifications

CAS Min % 97.0
Assay Percent Range 97%
Packaging Amber Glass Bottle
Quantity 1 g
Formula Weight 242.32
Percent Purity 97%
Chemical Name or Material 1-[4-(1H-Indol-3-yl)piperidino]ethan-1-one
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.