Learn More
1,3-O-Bis(triisopropylsilyl) 2-Oleoyl Glycerol-d5, TRC
Specifications
Specifications
| Chemical Name or Material | 1,3-O-Bis(triisopropylsilyl) 2-Oleoyl Glycerol-d5 |
| CAS | 1331658-79-7 |
| Molecular Formula | C39H75D5O4Si2 |
| Quantity | 5 mg |
| Synonym | (9Z)-9-Octadecenoic Acid 2-Triisopropylsilyloxy-1-(triisopropylsilyloxymethyl)ethyl-d5 Ester, 1,3-O-Bis(triisopropylsilyl)-2-monoolein-d5, 1,3-O-Bis(triisopropylsilyl)-2-monooleoylglycerol-d5, 1,3-O-Bis(triisopropylsilyl)-2-oleoylglycerol-d5, 1,3-O-Bis(triisopropylsilyl)-glycerol-d5 1,3-O-Bis(triisopropylsilyl)-2-monooleate, 1,3-O-Bis(triisopropylsilyl)-beta-monoolein-d5 |
| InChI Formula | InChI=1S/C39H80O4Si2/c1-14-15-16-17-18-19-20-21-22-23-24-25-26-27-28-29-39(40)43-38(30-41-44(32(2)3,33(4)5)34(6)7)31-42-45(35(8)9,36(10)11)37(12)13/h21-22,32-38H,14-20,23-31H2,1-13H3/b22-21-/i30D2,31D2,38D |
| SMILES | [2H]C([2H])(O[Si](C(C)C)(C(C)C)C(C)C)C([2H])(OC(=O)CCCCCCC\C=C/CCCCCCCC)C([2H])([2H])O[Si](C(C)C)(C(C)C)C(C)C |
| IUPAC Name | [1,1,2,3,3-pentadeuterio-1,3-bis[tri(propan-2-yl)silyloxy]propan-2-yl] (Z)-octadec-9-enoate |
| Molecular Weight (g/mol) | 674.25 |
| Formula Weight | 673.5909 |