missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,3-Indandione, TRC
Click to view available options
Quantity:
10 g
25 g
Unit Size:
10g
25g
Specifications
Specifications
| Chemical Name or Material | 1,3-Indanedione |
| CAS | 606-23-5 |
| Molecular Formula | C9 H6 O2 |
| Quantity | 25 g |
| Synonym | 1H-Indene-1,3(2H)-dione (9CI, ACI), 1,3-Indandione (8CI), Indandione (6CI), 1,3-Diketohydrindene, 1,3-Dioxobenzocyclopentane, 1,3-Indanedione, 2H-Indene-1,3-dione, Indene-1,3(2H)-dione, NSC 26329, NSC 6312 |
| InChI Formula | InChI=1S/C9H6O2/c10-8-5-9(11)7-4-2-1-3-6(7)8/h1-4H,5H2 |
| SMILES | O=C1CC(=O)c2ccccc12 |
| IUPAC Name | indene-1,3-dione |
| Molecular Weight (g/mol) | 146.14 |
| Formula Weight | 146.0368 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction