missing translation for 'onlineSavingsMsg'
Learn More

1,3-Distearoyl-2-chloropropanediol, TRC

Product Code. 17920105
Change view
Click to view available options
Quantity:
10 mg
25 mg
100 mg
Unit Size:
100mg
10mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17920105 10 mg 10mg
17998314 100 mg 100mg
17907454 25 mg 25mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 17920105 Supplier Toronto Research Chemicals Supplier No. TRCD49351010MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 26787-56-4
Molecular Formula C39 H75 Cl O4
Molecular Weight (g/mol) 643.46
Synonym Octadecanoic acid, 2-chloro-1,3-propanediyl ester (9CI), Stearic acid, 2-chlorotrimethylene ester (8CI), 1,3-Propanediol, 2-chloro-, distearate (8CI), Octadecanoic acid, 2-chloro-1,3-propanediyl ester (9CI), Stearic acid, 2-chlorotrimethylene ester (8CI), 1,3-Propanediol, 2-chloro-, distearate (8CI), 1,3-Bis-stearoyl-2-chloropropanediol
IUPAC Name (2-chloro-3-octadecanoyloxypropyl) octadecanoate
SMILES CCCCCCCCCCCCCCCCCC(=O)OCC(Cl)COC(=O)CCCCCCCCCCCCCCCCC

Specifications

Quantity 10 mg
InChI Formula InChI=1S/C39H75ClO4/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-31-33-38(41)43-35-37(40)36-44-39(42)34-32-30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h37H,3-36H2,1-2H3
Formula Weight 642.5354
Percent Purity >95%
Chemical Name or Material rac-1,3-Distearoyl-2-chloropropanediol
Grade HPLC
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.