Learn More
1,3-Dimethyl-5-nitroadamantane, TRC
Product Code. 30382634
Change view
Click to view available options
Quantity:
100 mg
500 mg
1 g
Unit Size:
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30382634 500 mg -
30370107 100 mg -
30372937 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30382634 Supplier Toronto Research Chemicals Supplier No. TRCD476665500MG
 Restricted Item
Estimated Shipment: 04-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6588-68-7
Molecular Formula C12H19NO2
Molecular Weight (g/mol) 209.28
Synonym 1,3-Dimethyl-5-nitroadamantane
IUPAC Name (1S,3R)-1,3-dimethyl-5-nitroadamantane
SMILES O=[N+]([O-])[C@@]1(C2)C[C@]3(C)C[C@H]2C[C@@](C1)(C)C3
Specifications
Quantity 500 mg
InChI Formula InChI=1S/C12H19NO2/c1-10-3-9-4-11(2,6-10)8-12(5-9,7-10)13(14)15/h9H,3-8H2,1-2H3/t9-,10+,11-,12-
Formula Weight 209.1416
Chemical Name or Material 1,3-Dimethyl-5-nitroadamantane