Learn More
1,3-Difluoropropane, 97%, Thermo Scientific™
Product Code. 11378755
Change view
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11378755 5 g 5g
11388755 25 g 25g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 11378755 Supplier Thermo Scientific Alfa Aesar Supplier No. L12351.06
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 462-39-5
Molecular Formula C3H6F2
Molecular Weight (g/mol) 80.08
MDL Number MFCD00040969
InChI Key OOLOYCGJRJFTPM-UHFFFAOYSA-N
Synonym propane, 1,3-difluoro, acmc-1aiey, 1,3-bis fluoranyl propane
PubChem CID 68041
IUPAC Name 1,3-difluoropropane
SMILES FCCCF
Specifications
Density 1.006
Boiling Point 40°C to 42°C
Linear Formula FCH2CH2CH2F
Refractive Index 1.321
Quantity 5 g
Beilstein 1696999
Formula Weight 80.08
Percent Purity 97%
Chemical Name or Material 1,3-Difluoropropane