Learn More
1,3-Di-HABA Kanamycin A, TRC
Product Code. 30012586
Change view
Click to view available options
Quantity:
1 mg
10 mg
25 mg
Unit Size:
10mg
1mg
25mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30012586 10 mg 10mg
30137841 25 mg 25mg
30011067 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30012586 Supplier Toronto Research Chemicals Supplier No. TRCD44637510MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 927821-99-6
Molecular Formula C26 H50 N6 O15
Molecular Weight (g/mol) 686.71
Synonym D-Streptamine, O-3-amino-3-deoxy-α-D-glucopyranosyl-(1→6)-O-[6-amino-6-deoxy-α-D-glucopyranosyl-(1→4)]-N1,N3-bis[(2S)-4-amino-2-hydroxy-1-oxobutyl]-2-deoxy-, O-3-Amino-3-deoxy-α-D-glucopyranosyl-(1→6)-O-[6-amino-6-deoxy-α-D-glucopyranosyl-(1→4)]-N1,N3-bis[(2S)-4-amino-2-hydroxy-1-oxobutyl]-2-deoxy-D-streptamine, 4-O-(3-Amino-3-deoxy-α-D-glucopyranosyl)-6-O-(6-amino-6-deoxy-α-D-glucopyranosyl)-1,3-N-bis[(2S)-4-amino-2-hydroxybutanoyl]-2-deoxy-L-streptamine, 1,3-Di-HABA Kanamycin A
IUPAC Name (2S)-4-amino-N-[(1R,2S,3R,4R,5S)-2-[(2S,3R,4S,5S,6R)-4-amino-3,5-dihydroxy-6-(hydroxymethyl)oxan-2-yl]oxy-5-[[(2S)-4-amino-2-hydroxybutanoyl]amino]-4-[(2R,3R,4S,5S,6R)-6-(aminomethyl)-3,4,5-trihydroxyoxan-2-yl]oxy-3-hydroxycyclohexyl]-2-hydroxybutanamide
SMILES NCC[C@H](O)C(=O)N[C@H]1C[C@@H](NC(=O)[C@@H](O)CCN)[C@H](O[C@H]2O[C@H](CO)[C@@H](O)[C@H](N)[C@H]2O)[C@@H](O)[C@@H]1O[C@H]3O[C@H](CN)[C@@H](O)[C@H](O)[C@H]3O
Specifications
Quantity 10 mg
InChI Formula InChI=1S/C26H50N6O15/c27-3-1-10(34)23(42)31-8-5-9(32-24(43)11(35)2-4-28)22(47-26-19(40)18(39)16(37)12(6-29)44-26)20(41)21(8)46-25-17(38)14(30)15(36)13(7-33)45-25/h8-22,25-26,33-41H,1-7,27-30H2,(H,31,42)(H,32,43)/t8-,9+,10+,11+,12-,13-,14+,15-,16-,17-,18+,19-,20-,21+,22-,25-,26-/m1/s1
Formula Weight 686.3334
Percent Purity >95%
Chemical Name or Material 1,3-Di-HABA Kanamycin A
Grade HPLC