missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzodioxole-N-phthalimido DL-threo-Droxidopa-d3, TRC

Product Code. 30335991
Change view
Click to view available options
Quantity:
0.5 mg
1 mg
2.5 mg
Unit Size:
0.5mg
1mg
2.50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30335991 1 mg 1mg
30338188 0.5 mg 0.5mg
30345294 2.5 mg 2.50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30335991 Supplier Toronto Research Chemicals Supplier No. TRCB2035011MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Benzodioxole-N-phthalimido DL-threo-Droxidopa-d3
Molecular Formula C18 D3 H10 N O7
Quantity 1 mg
InChI Formula InChI=1S/C18H13NO7/c20-15(9-5-6-12-13(7-9)26-8-25-12)14(18(23)24)19-16(21)10-3-1-2-4-11(10)17(19)22/h1-7,14-15,20H,8H2,(H,23,24)/t14-,15+/m0/s1/i5D,6D,7D
SMILES [2H]c1c([2H])c([C@@H](O)[C@H](N2C(=O)c3ccccc3C2=O)C(=O)O)c([2H])c4OCOc14
IUPAC Name (2S,3R)-2-(1,3-dioxoisoindol-2-yl)-3-hydroxy-3-(4,6,7-trideuterio-1,3-benzodioxol-5-yl)propanoic acid
Molecular Weight (g/mol) 358.317
Formula Weight 358.088
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.