missing translation for 'onlineSavingsMsg'
Learn More

1,3-Benzodioxole-N-phthalimido DL-threo-Droxidopa-13C2,15N, TRC

Product Code. 30344233
Change view
Click to view available options
Quantity:
50 mg
Unit Size:
50mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30344233 50 mg 50mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30344233 Supplier Toronto Research Chemicals Supplier No. TRCB20350250MG

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 1,3-Benzodioxole-N-phthalimido DL-threo-Droxidopa-13C2,15N
CAS 1330188-86-7
Molecular Formula C1613C2H1315NO7
Quantity 50 mg
Synonym (R,S)-rel-alpha-(1,3-Benzodioxol-5-ylhydroxymethyl)-1,3-dihydro-1,3-dioxo-2H-isoindole-2-acetic Acid-13C2,15N
InChI Formula InChI=1S/C18H13NO7/c20-15(9-5-6-12-13(7-9)26-8-25-12)14(18(23)24)19-16(21)10-3-1-2-4-11(10)17(19)22/h1-7,14-15,20H,8H2,(H,23,24)/t14-,15-/m1/s1/i14+1,18+1,19+1
SMILES O[C@@H]([13C@@H]([15N]1C(=O)c2ccccc2C1=O)[13C](=O)O)c3ccc4OCOc4c3
IUPAC Name (2R,3R)-3-(1,3-benzodioxol-5-yl)-2-(1,3-dioxoisoindol-2-yl)-3-hydroxy(1,2-^{13}C_{2})propanoic acid
Molecular Weight (g/mol) 358.28
Formula Weight 358.0729
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.