Learn More
1-(3,5-Dibromo-4-hydroxyphenyl)ethanone, 98%, BLD Pharm™
Specifications
Specifications
| CAS | 2887-72-1 |
| Molecular Formula | C8H6Br2O2 |
| MDL Number | MFCD00075779 |
| Quantity | 5 g |
| SMILES | CC(C1=CC(Br)=C(O)C(Br)=C1)=O |
| Molecular Weight (g/mol) | 293.94 |
| Percent Purity | 98% |
| Chemical Name or Material | 1-(3,5-Dibromo-4-hydroxyphenyl)ethanone |