Learn More
1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde, TRC
Specifications
Specifications
| Chemical Name or Material | 1-((2R,4S,5R)-4-Hydroxy-5-(hydroxymethyl)tetrahydrofuran-2-yl)-2,4-dioxo-1,2,3,4-tetrahydropyrimidine-5-carbaldehyde |
| CAS | 4494-26-2 |
| Molecular Formula | C10 H12 N2 O6 |
| Quantity | 2.5 mg |
| Synonym | alpha-Oxothymidine, 2'-Deoxy-5-formyluridine, 5-Formyl-2'-deoxyuridine, 5-Formyldeoxyuridine, NSC 148297 |
| InChI Formula | InChI=1S/C10H12N2O6/c13-3-5-2-12(10(17)11-9(5)16)8-1-6(15)7(4-14)18-8/h2-3,6-8,14-15H,1,4H2,(H,11,16,17)/t6-,7+,8+/m0/s1 |
| SMILES | OC[C@H]1O[C@H](C[C@@H]1O)N2C=C(C=O)C(=O)NC2=O |
| IUPAC Name | 1-[(2R,4S,5R)-4-hydroxy-5-(hydroxymethyl)oxolan-2-yl]-2,4-dioxopyrimidine-5-carbaldehyde |
| Molecular Weight (g/mol) | 256.21 |
| Formula Weight | 256.0695 |