Learn More
(±)-1,2-Propanediol-d8, CDN
Product Code. 30325483
Click to view available options
Quantity:
0.5 g
1 g
Unit Size:
0.50g
1g
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30325483
Brand: CDN CDND16561G
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Discover 4000+ high-quality deuterated compounds, shipping directly from stock for speed. We offer many unique API’s, including active pharmaceutical ingredients, nitrosoamines, amino acids, steroids, gases, pesticides, and fatty acids.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,2-Propanediol D8
CAS 80156-55-4
Molecular Formula C3 D8 O2
Quantity 1 g
Synonym 1,2-Propanediol D8, 1,2-Propanediol-d8, Propylene glycol-d8, 1,2-Propane-1,1,2,3,3,3-d6-diol-d2 (9 CI)
InChI Formula InChI=1 S/C3H8O2/c1-3(5)2-4/h3-5 H,2H2,1H3/i1D3,2D2,3 D,4 D,5 D
SMILES [2 H]OC([2 H])([2 H])C([2 H])(O[2 H])C([2 H])([2 H])[2 H]
IUPAC Name 1,1,1,2,3,3-hexadeuterio-2,3-dideuteriooxypropane
Molecular Weight (g/mol) 84.1437
Formula Weight 84.1026 g/mol
Percent Purity 98 atom % D, min 98% Chemical Purity