Learn More
1-(2-Furyl)ethanol-d3, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(2-Furyl)ethanol-d3 |
| CAS | 132331-94-3 |
| Molecular Formula | C6H5D3O2 |
| Quantity | 2.5 mg |
| Synonym | alpha-(Methyl-d3)-2-furanmethanol |
| InChI Formula | InChI=1S/C6H8O2/c1-5(7)6-3-2-4-8-6/h2-5,7H,1H3/i1D3 |
| SMILES | [2H]C([2H])([2H])C(O)c1occc1 |
| IUPAC Name | 2,2,2-trideuterio-1-(furan-2-yl)ethanol |
| Molecular Weight (g/mol) | 115.15 |
| Formula Weight | 115.0713 |