Learn More
1,2-Epoxy-9-decene, TRC
Specifications
Specifications
| Chemical Name or Material | 1,2-Epoxy-9-decene |
| CAS | 85721-25-1 |
| Molecular Formula | C10H18O |
| Quantity | 250 mg |
| InChI Formula | InChI=1S/C10H18O/c1-2-3-4-5-6-7-8-10-9-11-10/h2,10H,1,3-9H2 |
| SMILES | C=CCCCCCCC1CO1 |
| IUPAC Name | 2-oct-7-enyloxirane |
| Molecular Weight (g/mol) | 154.24 |
| Formula Weight | 154.1358 |