Learn More
1,2-Dipalmitoyl-sn-glycerol, TRC
Product Code. 30136420
Change view
Click to view available options
Quantity:
25 mg
100 mg
Unit Size:
100mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30136420 25 mg 25mg
17958684 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30136420 Supplier Toronto Research Chemicals Supplier No. TRCD48689025MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 30334-71-5
Molecular Formula C35H68O5
Molecular Weight (g/mol) 568.91
Synonym Hexadecanoic Acid 1,1'-[(1S)-1-(Hydroxymethyl)-1,2-ethanediyl] Ester, (S)-(-)-1,2-Dipalmitin, (S)-1-(Hydroxymethyl)ethane-1,2-diyl Dipalmitate, 1,2-Di-O-palmitoyl-sn-glycerol, L-1,2-Dipalmitin, NSC 269964, sn-1,2-Dipalmitin;
SMILES CCCCCCCCCCCCCCCC(=O)OC[C@H](CO)OC(=O)CCCCCCCCCCCCCCC
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C35H68O5/c1-3-5-7-9-11-13-15-17-19-21-23-25-27-29-34(37)39-32-33(31-36)40-35(38)30-28-26-24-22-20-18-16-14-12-10-8-6-4-2/h33,36H,3-32H2,1-2H3/t33-/m0/s1
Formula Weight 568.5067
Chemical Name or Material 1,2-Dipalmitoyl-sn-glycerol