missing translation for 'onlineSavingsMsg'
Learn More

1,2-Diformylhydrazine, TRC

Product Code. 30379620
Change view
Click to view available options
Quantity:
25 g
100 g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30379620 25 g
30384012 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 30379620 Supplier Toronto Research Chemicals Supplier No. TRCD44732025G
 Restricted Item
Estimated Shipment: 27-07-2026
Add to basket
Add to basket
This item is not returnable. View return policy

High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 628-36-4
Molecular Formula C2H4N2O2
Molecular Weight (g/mol) 88.07
Synonym 1,2-Hydrazinedicarboxaldehyde, Bicarbamaldehyde, Diformohydrazide, Diformylhydrazide, Diformylhydrazine, N,N'-Diformylhydrazine, NSC 54729, s-Diformohydrazide, s-Diformylhydrazine
SMILES O=CNNC=O

Specifications

Quantity 25 g
InChI Formula InChI=1S/C2H4N2O2/c5-1-3-4-2-6/h1-2H,(H,3,5)(H,4,6)
Chemical Name or Material 1,2-Diformylhydrazine
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.