Learn More
1-(2-Chlorophenyl)-1,4,6,7-tetrahydro-4-(3-methylphenyl)-7-oxo-3-phenyl-8H-pyrazolo[3,4-e][1,4]thiazepine-8-acetic Acid, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(2-Chlorophenyl)-1,4,6,7-tetrahydro-4-(3-methylphenyl)-7-oxo-3-phenyl-8H-pyrazolo[3,4-e][1,4]thiazepine-8-acetic Acid |
| CAS | 1043712-19-1 |
| Molecular Formula | C27H22ClN3O3S |
| Quantity | 25 mg |
| InChI Formula | InChI=1S/C27H22ClN3O3S/c1-17-8-7-11-19(14-17)26-24-25(18-9-3-2-4-10-18)29-31(21-13-6-5-12-20(21)28)27(24)30(15-23(33)34)22(32)16-35-26/h2-14,26H,15-16H2,1H3,(H,33,34) |
| SMILES | Cc1cccc(c1)C2SCC(=O)N(CC(=O)O)c3c2c(nn3c4ccccc4Cl)c5ccccc5 |
| IUPAC Name | 2-[1-(2-chlorophenyl)-4-(3-methylphenyl)-7-oxo-3-phenyl-4H-pyrazolo[3,4-e][1,4]thiazepin-8-yl]acetic acid |
| Molecular Weight (g/mol) | 504 |
| Formula Weight | 503.107 |