Learn More
1-(2-Chloro-5-nitrophenyl)-2,5-dihydro-1H-pyrrole-2,5-dione (~85%), TRC
Product Code. 30367508
Change view
Click to view available options
Quantity:
50 mg
100 mg
500 mg
Unit Size:
100mg
500mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30367508 100 mg 100mg
30349046 50 mg 50mg
30349887 500 mg 500mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30367508 Supplier Toronto Research Chemicals Supplier No. TRCC613690100MG
 Restricted Item
Estimated Shipment: 12-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 313069-92-0
Molecular Formula C10H5N2O4Cl
Molecular Weight (g/mol) 252.61
Synonym 1H-Pyrrole-2,5-dione, 1-(2-chloro-5-nitrophenyl)- (9CI, ACI), 1-(2-Chloro-5-nitrophenyl)-1H-pyrrole-2,5-dione (ACI), N-(2-Chloro-5-nitrophenyl)maleimide
IUPAC Name 1-(2-chloro-5-nitrophenyl)pyrrole-2,5-dione
SMILES [O-][N+](=O)c1ccc(Cl)c(c1)N2C(=O)C=CC2=O
Specifications
Quantity 100 mg
InChI Formula InChI=1S/C10H5ClN2O4/c11-7-2-1-6(13(16)17)5-8(7)12-9(14)3-4-10(12)15/h1-5H
Formula Weight 251.9938
Chemical Name or Material 1-(2-Chloro-5-nitrophenyl)-2,5-dihydro-1H-pyrrole-2,5-dione