Learn More
1-(2-Bromophenyl)piperazine, TRC
Product Code. 30347576
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347576 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30347576 Supplier Toronto Research Chemicals Supplier No. TRCB802705100MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1-(2-Bromophenyl)piperazine
CAS 1011-13-8
Molecular Formula C10 H13 Br N2
Quantity 100 mg
InChI Formula InChI=1S/C10H13BrN2/c11-9-3-1-2-4-10(9)13-7-5-12-6-8-13/h1-4,12H,5-8H2
SMILES Brc1ccccc1N2CCNCC2
IUPAC Name 1-(2-bromophenyl)piperazine
Molecular Weight (g/mol) 241.128
Formula Weight 240.026