Learn More
1,2-Bis(2-aminophenoxy)ethane, TRC
Specifications
Specifications
| Chemical Name or Material | 1,2-Bis(2-aminophenoxy)ethane |
| CAS | 52411-34-4 |
| Molecular Formula | C14 H16 N2 O2 |
| Quantity | 2.5 g |
| InChI Formula | InChI=1S/C14H16N2O2/c15-11-5-1-3-7-13(11)17-9-10-18-14-8-4-2-6-12(14)16/h1-8H,9-10,15-16H2 |
| SMILES | Nc1ccccc1OCCOc2ccccc2N |
| IUPAC Name | 2-[2-(2-aminophenoxy)ethoxy]aniline |
| Molecular Weight (g/mol) | 244.29 |
| Formula Weight | 244.1212 |