Learn More
1,2,6-Trimethyl-phenanthrene, TRC
Product Code. 30134429
Change view
Click to view available options
Quantity:
2.5 mg
25 mg
Unit Size:
2.5mg
25mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30134429 25 mg 25mg
30008511 2.5 mg 2.5mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 30134429 Supplier Toronto Research Chemicals Supplier No. TRCT79661025MG
 Restricted Item
Estimated Shipment: 11-11-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 30436-55-6
Molecular Formula C17 H16
Molecular Weight (g/mol) 220.31
Synonym 1,2,6-Trimethyl-phenanthrene, 1,2,6-Trimethylphenanthrene
IUPAC Name 1,2,6-trimethylphenanthrene
SMILES Cc1ccc2ccc3c(C)c(C)ccc3c2c1
Specifications
Quantity 25 mg
InChI Formula InChI=1S/C17H16/c1-11-4-6-14-7-9-15-13(3)12(2)5-8-16(15)17(14)10-11/h4-10H,1-3H3
Formula Weight 220.1252
Chemical Name or Material 1,2,6-Trimethylphenanthrene