Learn More
1,2,4-Triazole-d3, CDN
Click to view available options
Quantity:
0.25 g
0.5 g
Unit Size:
0.25g
0.5g
Specifications
Specifications
| Chemical Name or Material | 1,2,4-Triazole-d3 |
| CAS | 43088-92-2 |
| Molecular Formula | C2 D3 N3 |
| Quantity | 0.25 g |
| InChI Formula | InChI=1 S/C2H3N3/c1-3-2-5-4-1/h1-2 H,(H,3,4,5)/i1D,2 D/hD |
| SMILES | [2 H]c1nc([2 H])n([2 H])n1 |
| IUPAC Name | 1,3,5-trideuterio-1,2,4-triazole |
| Molecular Weight (g/mol) | 72.084 |
| Formula Weight | 72.052 g/mol |
| Percent Purity | 98 atom % D, min 98% Chemical Purity |