missing translation for 'onlineSavingsMsg'
Learn More

1,2,3-Trifluoro-4-nitrobenzene, 97%

Product Code. 11487777
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
Product Code. Quantity unitSize
11487777 5 g 5g
11497777 25 g 25g
11407787 100 g 100g
3 options
This item is not returnable. View return policy

Product Code. 11487777

Brand: Thermo Scientific Alfa Aesar A16313.06

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 771-69-7 | C6H2F3NO2 | 177.08 g/mol

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 771-69-7
Molecular Formula C6H2F3NO2
Molecular Weight (g/mol) 177.08
MDL Number MFCD00041546
InChI Key ARCACZWMYGILNI-UHFFFAOYSA-N
Synonym 2,3,4-trifluoronitrobenzene, 2,3,4-trifluoro-1-nitrobenzene, 4-nitro-1,2,3-trifluorobenzene, trifluoronitrobenzene, benzene, 1,2,3-trifluoro-4-nitro, 2,3,4-trifluornitrobenzene, 1,2,3-trifluoro-4-nitro-benzene, 1-nitro-2,3,4-trifluorobenzene, pubchem2287, benzene, trifluoronitro
PubChem CID 69871
IUPAC Name 1,2,3-trifluoro-4-nitrobenzene
SMILES [O-][N+](=O)C1=CC=C(F)C(F)=C1F

Specifications

Density 1.541
Boiling Point 91°C to 93°C (20 mmHg)
Flash Point 93°C (199°F)
Refractive Index 1.492
Quantity 5 g
UN Number UN2810
Beilstein 2449746
Formula Weight 177.08
Percent Purity 97%
Chemical Name or Material 1,2,3-Trifluoro-4-nitrobenzene

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.