Learn More
1-(2,3-Epoxypropyl)-2-nitroimidazole, TRC
Specifications
Specifications
| Chemical Name or Material | 1-(2,3-Epoxypropyl)-2-nitroimidazole |
| CAS | 13551-90-1 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C6H7N3O3 |
| Quantity | 250 mg |
| Synonym | 2-Nitro-1-(oxiranylmethyl)-1H-imidazole, 1-(2,3-Epoxypropyl)-2-nitro-imidazole, 2-Nitro-1-(2-oxiranylmethyl)-1H-imidazole, 1-(Oxiranylmethyl)-2-nitro-1H-imidazole, 1-Oxiranylmethyl-2-nitroimidazole, NSC 342688 |
| InChI Formula | InChI=1S/C6H7N3O3/c10-9(11)6-7-1-2-8(6)3-5-4-12-5/h1-2,5H,3-4H2 |
| SMILES | [O-][N+](=O)c1nccn1CC2CO2 |
| IUPAC Name | 2-nitro-1-(oxiran-2-ylmethyl)imidazole |
| Molecular Weight (g/mol) | 169.14 |
| Show More |