Learn More
1-(2,3,5,6-Tetramethylphenylsulfonylamino)cyclohexanecarboxylic acid, 95%, Thermo Scientific™
Product Code. 15497925
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
Product Code. 15497925
Brand: Thermo Scientific Alfa Aesar H31898.03
Request Bulk or Custom Format
This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 885269-54-5
Molecular Formula C17H25NO4S
Molecular Weight (g/mol) 339.45
MDL Number MFCD06409473
InChI Key AUJXKRTUDQFEGI-UHFFFAOYSA-N
Synonym 1-2,3,5,6-tetramethylphenyl sulfonamido cyclohexane-1-carboxylic acid, 1-2,3,5,6-tetramethylphenylsulfonamido cyclohexanecarboxylic acid, 1-2,3,5,6-tetramethylphenylsulfonylamino cyclohexanecarboxylic acid, 1-2,3,5,6-tetramethylbenzenesulfonamido cyclohexane-1-carboxylic acid, 1-2,3,5,6-tetramethylbenzene-1-sulfonyl amino cyclohexane-1-carboxylic acid
PubChem CID 44669178
IUPAC Name 1-[(2,3,5,6-tetramethylphenyl)sulfonylamino]cyclohexane-1-carboxylic acid
SMILES CC1=CC(=C(C(=C1C)S(=O)(=O)NC2(CCCCC2)C(=O)O)C)C
Specifications
Melting Point ∽212°C (decomposition)
Quantity 1 g
Formula Weight 339.46
Percent Purity 95%
Chemical Name or Material 1-(2,3,5,6-Tetramethylphenylsulfonylamino)cyclohexanecarboxylic acid