Learn More
1,2,3,4-Tetra-O-acetyl-Alpha,Beta-D-glucuronic Acid, Methyl Ester (mixture of anomers), TRC
Specifications
Specifications
| Chemical Name or Material | 1,2,3,4-Tetra-O-acetyl-alpha,beta-D-glucuronic Acid','Methyl Ester (mixture of anomers) |
| CAS | 3082-96-0 |
| Molecular Formula | C15H20O11 |
| Quantity | 2.5 g |
| Synonym | Methyl 1,2,3,4-Tetra-O-Acetyl-alpha,beta-D-glucopyranosiduronate, D-Glucopyranuronic Acid Methyl Ester Tetraacetate |
| InChI Formula | InChI=1S/C15H20O11/c1-6(16)22-10-11(23-7(2)17)13(24-8(3)18)15(25-9(4)19)26-12(10)14(20)21-5/h10-13,15H,1-5H3/t10-,11-,12-,13+,15?/m0/s1 |
| SMILES | COC(=O)[C@H]1OC(OC(=O)C)[C@H](OC(=O)C)[C@@H](OC(=O)C)[C@@H]1OC(=O)C |
| IUPAC Name | methyl (2S,3S,4S,5R)-3,4,5,6-tetraacetyloxyoxane-2-carboxylate |
| Molecular Weight (g/mol) | 376.31 |
| Formula Weight | 376.1006 |