missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,12-Dodecane-d24-diol, CDN
Click to view available options
Quantity:
0.05 g
0.1 g
Unit Size:
0.1g
Specifications
Specifications
| Chemical Name or Material | 1,12-Dodecanediol-D24 |
| CAS | 1219802-88-6 |
| Molecular Formula | C12 D24 H2 O2 |
| Quantity | 0.1 g |
| Synonym | 1,12-Dodecanediol-D24, 1,12-Dihydroxydodecane-1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-d24, α,ω-Dodecanediol-d24, 1,12-Dodecane-d24-diol |
| InChI Formula | InChI=1 S/C12H26O2/c13-11-9-7-5-3-1-2-4-6-8-10-12-14/h13-14 H,1-12H2/i1D2,2D2,3D2,4D2,5D2,6D2,7D2,8D2,9D2,10D2,11D2,12D2 |
| SMILES | [2 H]C([2 H])(O)C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])C([2 H])([2 H])O |
| IUPAC Name | 1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-tetracosadeuteriododecane-1,12-diol |
| Molecular Weight (g/mol) | 226.4815 |
| Formula Weight | 226.3439 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction