Learn More
1,12-Dibromododecane, 98%
Product Code. 11484777
Change view
Click to view available options
Quantity:
5 g
25 g
100 g
Unit Size:
100g
25g
5g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
11484777 5 g 5g
11494777 25 g 25g
11404787 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 11484777 Supplier Thermo Scientific Alfa Aesar Supplier No. B21254.06
 Restricted Item
In Stock
Add to basket
CAS: 3344-70-5 | C12H24Br2 | 328.132 g/mol
1,12-Dibromododecane, is used as an organic chemical intermediate.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,12-Dibromododecane, is used as an organic chemical intermediate.
Solubility
Soluble in hot methanol- almost transparency.
Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Stable under recommended storage conditions.Stable. Combustible. Incompatible with strong oxidizing agents.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 3344-70-5
Molecular Formula C12H24Br2
Molecular Weight (g/mol) 328.132
MDL Number MFCD00000226
InChI Key ZJJATABWMGVVRZ-UHFFFAOYSA-N
Synonym dodecamethylene dibromide, dodecane, 1,12-dibromo, alpha,omega-dibromododecane, labotest-bb ltbb002671, pubchem3879, 1, 12-dibromododecane, 1,12-dibromo-dodecane, 1,12-bis bromanyl dodecane, 4-01-00-00503 beilstein handbook reference, ksc225m2h
PubChem CID 18766
IUPAC Name 1,12-dibromododecane
SMILES C(CCCCCCBr)CCCCCBr
Specifications
Melting Point 38°C to 42°C
Boiling Point 215°C (15 mmHg)
Flash Point >110°C (230°F)
Quantity 5 g
Beilstein 1742763
Solubility Information Soluble in hot methanol- almost transparency.
Formula Weight 328.13
Percent Purity 98%
Chemical Name or Material 1,12-Dibromododecane