missing translation for 'onlineSavingsMsg'
Learn More
Learn More
1,1-Dichloro-2,2-bis(4-chlorophenyl-d4)ethylene, CDN
Click to view available options
Quantity:
0.01 g
Unit Size:
0.01g
Specifications
Specifications
| Chemical Name or Material | 4,4'-DDED8 |
| CAS | 93952-19-3 |
| Molecular Formula | C14 2H8 Cl4 |
| Quantity | 0.01 g |
| Synonym | Benzene-1,2,4,5-d4, 3,3'-(dichloroethenylidene)bis[6-chloro- (9 CI), 4,4'-DDE D8 |
| InChI Formula | InChI=1 S/C14H8Cl4/c15-11-5-1-9(2-6-11)13(14(17)18)10-3-7-12(16)8-4-10/h1-8 H/i1D,2 D,3 D,4 D,5 D,6 D,7 D,8 D |
| SMILES | [2 H]c1c([2 H])c(C(=C(Cl)Cl)c2c([2 H])c([2 H])c(Cl)c([2 H])c2[2 H])c([2 H])c([2 H])c1Cl |
| IUPAC Name | 1-chloro-2,3,5,6-tetradeuterio-4-[2,2-dichloro-1-(4-chloro-2,3,5,6-tetradeuteriophenyl)ethenyl]benzene |
| Molecular Weight (g/mol) | 326.07 |
| Formula Weight | 323.9882 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction