missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate, 97%

Product Code. 15425295
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
This item is not returnable. View return policy

Product Code. 15425295

Brand: Thermo Scientific Alfa Aesar H55957.ME

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 41085-99-8 | C59H98ClN2O4+ | 934.893 g/mol

1,1'-Dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine per chlorate is a lipophilic dye used to record membrane voltage for fluidity studies. This retrograde stain is useful in staining neurons because the stain is relatively nontoxic to cell bodies and the dye is retained in neurons. In addition, 1,1'-Dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine per chlorate is an ideal retrograde and anterograde tracer to label the processes as well as the cell bodies of cultured neurons.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine per chlorate is a lipophilic dye used to record membrane voltage for fluidity studies. This retrograde stain is useful in staining neurons because the stain is relatively nontoxic to cell bodies and the dye is retained in neurons. In addition, 1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine per chlorate is an ideal retrograde and anterograde tracer to label the processes as well as the cell bodies of cultured neurons.

Solubility
Soluble in Dimethylformamide (DMF).

Notes
Reacts with strong oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 41085-99-8
Molecular Formula C59H98ClN2O4+
Molecular Weight (g/mol) 934.893
MDL Number MFCD00142354
InChI Key JVXZRNYCRFIEGV-UHFFFAOYSA-N
Synonym 1,1'-dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate
PubChem CID 129893813
IUPAC Name (2E)-2-[(E)-3-(3,3-dimethyl-1-octadecylindol-1-ium-2-yl)prop-2-enylidene]-3,3-dimethyl-1-octadecylindole;perchloric acid
SMILES CCCCCCCCCCCCCCCCCCN1C2=CC=CC=C2C(C1=CC=CC3=[N+](C4=CC=CC=C4C3(C)C)CCCCCCCCCCCCCCCCCC)(C)C.OCl(=O)(=O)=O

Specifications

Melting Point 68°C (decomposition)
Quantity 500 mg
Beilstein 8251870
Solubility Information Soluble in Dimethylformamide (DMF).
Formula Weight 933.88
Percent Purity 97%
Chemical Name or Material 1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.