Learn More
1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate, 97%
Product Code. 15415295
Click to view available options
Quantity:
100 mg
500 mg
Unit Size:
100mg
500mg
Product Code. 15415295
Brand: Thermo Scientific Alfa Aesar H55957.MC
Request Bulk or Custom Format
CAS: 41085-99-8 | C59H98ClN2O4+ | 934.893 g/mol
1,1'-Dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine per chlorate is a lipophilic dye used to record membrane voltage for fluidity studies. This retrograde stain is useful in staining neurons because the stain is relatively nontoxic to cell bodies and the dye is retained in neurons. In addition, 1,1'-Dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine per chlorate is an ideal retrograde and anterograde tracer to label the processes as well as the cell bodies of cultured neurons.
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Applications
1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine per chlorate is a lipophilic dye used to record membrane voltage for fluidity studies. This retrograde stain is useful in staining neurons because the stain is relatively nontoxic to cell bodies and the dye is retained in neurons. In addition, 1,1′-Dioctadecyl-3,3,3′,3′-tetramethylindocarbocyanine per chlorate is an ideal retrograde and anterograde tracer to label the processes as well as the cell bodies of cultured neurons.
Solubility
Soluble in Dimethylformamide (DMF).
Notes
Reacts with strong oxidizing agents. Store at room temperature.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 41085-99-8
Molecular Formula C59H98ClN2O4+
Molecular Weight (g/mol) 934.893
MDL Number MFCD00142354
InChI Key JVXZRNYCRFIEGV-UHFFFAOYSA-N
Synonym 1,1'-dioctadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate
PubChem CID 129893813
IUPAC Name (2E)-2-[(E)-3-(3,3-dimethyl-1-octadecylindol-1-ium-2-yl)prop-2-enylidene]-3,3-dimethyl-1-octadecylindole;perchloric acid
SMILES CCCCCCCCCCCCCCCCCCN1C2=CC=CC=C2C(C1=CC=CC3=[N+](C4=CC=CC=C4C3(C)C)CCCCCCCCCCCCCCCCCC)(C)C.OCl(=O)(=O)=O
Specifications
Melting Point 68°C (decomposition)
Quantity 100 mg
Beilstein 8251870
Solubility Information Soluble in Dimethylformamide (DMF).
Formula Weight 933.88
Percent Purity 97%
Chemical Name or Material 1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate
compliance-icons
Product Identifier
  • 1,1'-Di-n-octadecyl-3,3,3',3'-tetramethylindocarbocyanine perchlorate
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
  • P332+P313-If skin irritation occurs: Get medical advice/attention.
  • P337+P313-If eye irritation persists: Get medical advice/attention.
Supplemental information
  • MIXTURE LIST-Contains : 3H-Indolium, 2-[3-(1,3-dihydro-3,3-dimethyl-1-octadecyl2H-indol-2-ylidene)-1-propen-1-yl]-3,3-dimeth