Learn More
1,1'-(Decane-1,10-diyl)bis(N-octylpyridin-4(1H)-imine) dihydrochloride, 98%, BLD Pharm™
Product Code. 31428744
Encompass_Preferred
Change view
Click to view available options
Quantity:
250 mg
1 g
5 g
10 g
25 g
100 g
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
31428744 1 g
31427090 25 g
31419770 5 g
31418061 250 mg
31420414 10 g
31428037 100 g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 31428744 Supplier BLD Pharm Supplier No. BD1417351g
 Restricted Item
Estimated Shipment: 25-11-2026
Specifications
CAS 70775-75-6
Molecular Formula C36H64Cl2N4
MDL Number MFCD01938808
Quantity 1 g
SMILES CCCCCCCC/N=C1C=CN(CCCCCCCCCCN(C=C/2)C=CC2=N/CCCCCCCC)C=C/1.[H]Cl.[H]Cl
Molecular Weight (g/mol) 623.83
Percent Purity 98%
Chemical Name or Material 1,1'-(Decane-1,10-diyl)bis(N-octylpyridin-4(1H)-imine) dihydrochloride