Learn More
1,1-Cyclopentanediacetic Anhydride, TRC
Specifications
Specifications
| Chemical Name or Material | 1,1-Cyclopentanediacetic Anhydride |
| CAS | 5662-95-3 |
| Molecular Formula | C9H12O3 |
| Quantity | 25 g |
| Synonym | 1,1-Cyclopentanediacetic Acid Anhydride, 3,3-Tetramethyleneglutaric Anhydride, NSC 78448 |
| InChI Formula | InChI=1S/C9H12O3/c10-7-5-9(3-1-2-4-9)6-8(11)12-7/h1-6H2 |
| SMILES | O=C(O1)CC2(CCCC2)CC1=O |
| IUPAC Name | 8-oxaspiro[4.5]decane-7,9-dione |
| Molecular Weight (g/mol) | 168.19 |
| Formula Weight | 168.08 |