Learn More
1,1-Bis(p-isobutylphenyl)ethane, TRC
Product Code. 30347451
Change view
Click to view available options
Quantity:
100 mg
Unit Size:
100mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30347451 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30347451 Supplier Toronto Research Chemicals Supplier No. TRCB448875100MG
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,1'-(Ethane-1,1-diyl)-4,4'-(2-methylpropyl)dibenzene
CAS 102120-87-6
Molecular Formula C22 H30
Quantity 100 mg
Synonym 1,1'-(Ethane-1,1-diyl)-4,4'-(2-methylpropyl)dibenzene, Ibuprofen Imp. R (EP)
InChI Formula InChI=1S/C22H30/c1-16(2)14-19-6-10-21(11-7-19)18(5)22-12-8-20(9-13-22)15-17(3)4/h6-13,16-18H,14-15H2,1-5H3
SMILES CC(C)Cc1ccc(cc1)C(C)c2ccc(CC(C)C)cc2
IUPAC Name 1-(2-methylpropyl)-4-[1-[4-(2-methylpropyl)phenyl]ethyl]benzene
Molecular Weight (g/mol) 294.47
Formula Weight 294.2348