Learn More
1,1-Bis(p-isobutylphenyl)ethane, TRC
Product Code. 30344866
Change view
Click to view available options
Quantity:
1 g
Unit Size:
1g
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30344866 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 30344866 Supplier Toronto Research Chemicals Supplier No. TRCB4488751G
 Restricted Item
Estimated Shipment: 14-12-2026
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 1,1'-(Ethane-1,1-diyl)-4,4'-(2-methylpropyl)dibenzene
CAS 102120-87-6
Molecular Formula C22 H30
Quantity 1 g
Synonym 1,1'-(Ethane-1,1-diyl)-4,4'-(2-methylpropyl)dibenzene, Ibuprofen Imp. R (EP)
InChI Formula InChI=1S/C22H30/c1-16(2)14-19-6-10-21(11-7-19)18(5)22-12-8-20(9-13-22)15-17(3)4/h6-13,16-18H,14-15H2,1-5H3
SMILES CC(C)Cc1ccc(cc1)C(C)c2ccc(CC(C)C)cc2
IUPAC Name 1-(2-methylpropyl)-4-[1-[4-(2-methylpropyl)phenyl]ethyl]benzene
Molecular Weight (g/mol) 294.47
Formula Weight 294.2348