missing translation for 'onlineSavingsMsg'
Learn More

1,1'-Biadamantane, Thermo Scientific Chemicals

Product Code. 16687052 Shop All Thermo Scientific Chemicals Products
Change view
Click to view available options
Quantity:
250 mg
1 g
Unit Size:
1g
250mg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16687052 250 mg 250mg
16677052 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 16687052 Supplier Thermo Scientific Acros Supplier No. 465932500
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 3732-31-8
Molecular Formula C20H30
Molecular Weight (g/mol) 270.46
MDL Number MFCD01910632
InChI Key MPXKIFWZOQVOLN-UHFFFAOYSA-N
Synonym 1,1 '-Diadamantane; Bisadamantane
IUPAC Name 1,1'-biadamantane
SMILES C1C2CC3CC1CC(C2)(C3)C12CC3CC(CC(C3)C1)C2

Specifications

Alpha Vector BIADAMANTANE
Melting Point 285°C to 295°C
Color White
Infrared Spectrum Conforms
Packaging glass bottle
Quantity 250 mg
Formula Weight 270.46
Physical Form Powder
Chemical Name or Material 1,1 '-Biadamantane

RUO – Research Use Only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.