Learn More
1,1,3,3-Tetraethoxypropane-1,3-d2, TRC
Specifications
Specifications
| Chemical Name or Material | 1,1,3,3-Tetraethoxypropane-1,3-d2 |
| CAS | 105479-86-5 |
| Purity Grade Notes | GC |
| Molecular Formula | C11 2H2 H22 O4 |
| Quantity | 5 mg |
| Synonym | Propane-1,3-d2','1,1,3,3-tetraethoxy- (9CI), Malonaldehyde-1,3-d2 Bis(diethyl Acetal), 1,1,3,3-Tetraethoxypropane-1,3-d2, 1,1,3,3-Tetraethoxypropane-1,3-D2 |
| InChI Formula | InChI=1S/C11H24O4/c1-5-12-10(13-6-2)9-11(14-7-3)15-8-4/h10-11H,5-9H2,1-4H3/i10D,11D |
| SMILES | [2H]C(CC([2H])(OCC)OCC)(OCC)OCC |
| IUPAC Name | 1,3-dideuterio-1,1,3,3-tetraethoxypropane |
| Molecular Weight (g/mol) | 222.32 |
| Show More |