missing translation for 'onlineSavingsMsg'
Learn More

1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione, 95%

Product Code. 11469352
Click to view available options
Quantity:
5 g
25 g
Unit Size:
25g
5g
This item is not returnable. View return policy

Product Code. 11469352

Brand: Thermo Scientific Alfa Aesar A10112.14

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item has been discontinued and is no longer available. View the product for possible alternatives or contact our Technical Support team on +351 21 425 33 50 for assistance
View alternative products

This item is not returnable. View return policy

CAS: 22767-90-4 | C8H11F3O2 | 196.17 g/mol

Useful for solvent extraction and GC separation of metals.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Useful for solvent extraction and GC separation of metals.

Notes
Store in cool, dry conditions in well sealed container. Store away from oxidizing agent.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 22767-90-4
Molecular Formula C8H11F3O2
Molecular Weight (g/mol) 196.17
MDL Number MFCD00040841
InChI Key BVPKYBMUQDZTJH-UHFFFAOYSA-N
Synonym 1,1,1-trifluoro-5,5-dimethyl-2,4-hexanedione, pivaloyltrifluoroacetone, 2,4-hexanedione, 1,1,1-trifluoro-5,5-dimethyl, 5,5-dimethyl-1,1,1-trifluorohexane-2,4-dione, 2,4-hexanedione,1,1,1-trifluoro-5,5-dimethyl, acmc-1cann, tos-bb-0655, 1,1,1-trifluoro-5,5,-dimethyl-2,4-hexanedione
PubChem CID 89830
IUPAC Name 1,1,1-trifluoro-5,5-dimethylhexane-2,4-dione
SMILES CC(C)(C)C(=O)CC(=O)C(F)(F)F

Specifications

Density 1.13
Boiling Point 68°C to 71°C (6 mmHg)
Flash Point 34°C (93°F)
Refractive Index 1.406
Quantity 25 g
UN Number UN1224
Beilstein 1708830
Formula Weight 196.17
Percent Purity 95%
Chemical Name or Material 1,1,1-Trifluoro-5,5-dimethyl-2,4-hexanedione

RUO – Research Use Only

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.