Learn More
1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-Pentacosafluoro-14-iodotetradecane, TRC
Specifications
Specifications
| Chemical Name or Material | 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-Pentacosafluoro-14-iodotetradecane |
| CAS | 30046-31-2 |
| Purity Grade Notes | GC |
| Molecular Formula | C14 H4 F25 I |
| Quantity | 100 mg |
| Synonym | 1H,1H,2H,2H-Perfluoro-1-iodotetradecane, 1H,1H,2H,2H-Perfluoro Iodotetradecane, 13H,13H,14H,14H-Perfluoro-14-iodotetradecane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-Pentacosafluoro-14-iodotetradecane (ACI), 1,1,2,2-Tetrahydroperfluorotetradecyl iodide, 1H,1H,2H,2H-Perfluorotetradecyl iodide, Perfluorolaurylethyl iodide, Tetradecane','1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-pentacosafluoro-14-iodo- (8CI','9CI','ACI), 1H,1H,2H,2H-Perfluorotetradecyl Iodide, 1H,1H,2H,2H-Perfluoro-1-iodotetradecane, 1H,1H,2H,2H-Perfluoro Iodotetradecane, 13H,13H,14H,14H-Perfluoro-14-iodotetradecane, 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-Pentacosafluoro-14-iodotetradecane (ACI), 1,1,2,2-Tetrahydroperfluorotetradecyl iodide, 1H,1H,2H,2H-Perfluorotetradecyl iodide, Perfluorolaurylethyl iodide, Tetradecane','1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-pentacosafluoro-14-iodo- (8CI','9CI','ACI), 1H,1H,2H,2H-Perfluorotetradecyl Iodide |
| InChI Formula | InChI=1S/C14H4F25I/c15-3(16,1-2-40)4(17,18)5(19,20)6(21,22)7(23,24)8(25,26)9(27,28)10(29,30)11(31,32)12(33,34)13(35,36)14(37,38)39/h1-2H2 |
| SMILES | FC(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)C(F)(F)CCI |
| IUPAC Name | 1,1,1,2,2,3,3,4,4,5,5,6,6,7,7,8,8,9,9,10,10,11,11,12,12-pentacosafluoro-14-iodotetradecane |
| Molecular Weight (g/mol) | 774.046 |
| Show More |