missing translation for 'onlineSavingsMsg'
Learn More
Learn More
alpha-Zearalenol-d7, TRC
Click to view available options
Quantity:
0.5 mg
5 mg
Unit Size:
0.5mg
5mg
Specifications
Specifications
| Chemical Name or Material | alpha-Zearalenol-d7 |
| Purity Grade Notes | HPLC |
| Molecular Formula | C18H17D7O5 |
| Quantity | 0.5 mg |
| Synonym | (3S,7R,11E)-3,4,5,6,7,8,9,10-Octahydro-7,14,16-trihydroxy-3-methyl-1H-2-benzoxacyclotetradecin-1-one-d7, (-)-α-Zearalenol-d7, Zearalenol-d7, trans-Zearalenol-d7 |
| SMILES | O=C1O[C@@H](C)CCC([2H])([2H])[C@]([2H])(O)C([2H])([2H])CC/C=C/C2=C([2H])C(O)=C([2H])C(O)=C21 |
| Molecular Weight (g/mol) | 327.42 |
| Percent Purity | >95 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction